6-Bromo-4-nitro-1H-indazole 97%
Supplier: Apollo Scientific
Warning
OR61053-5G
APOSOR61053-5GEA 116 EUR
APOSOR61053-5G
6-Bromo-4-nitro-1H-indazole 97%
6-Bromo-4-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
Melting Pt: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD07368216
CAS Number: 885518-46-7