2-Hydrazinylpyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2(1H)-Pyridinone, hydrazone, 2-hydrazinylpyridine, 2-Hydrazinopyridine
OR61068-100G OR61068-25G OR61068-5G
APOSOR61068-100GEA 478 EUR
APOSOR61068-100G APOSOR61068-25G APOSOR61068-5G
2-Hydrazinylpyridine 95%
2-Hydrazinylpyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 90…92 °C/0.75mmHg
Melting Pt: 41…44 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006249
CAS Number: 4930-98-7