3,5-Xylidine 97%
Supplier: Apollo Scientific
Danger
OR61129-100G OR61129-25G OR61129-5G
APOSOR61129-100GEA 71.2 EUR
APOSOR61129-100G APOSOR61129-25G APOSOR61129-5G
3,5-Xylidine 97%
3,5-Xylidine
Formula: C₈H₁₁N
Boiling Pt: 221 °C
Melting Pt: 44446 °C
Density: 0.972 g/cm³ (25 °C)
Flash Pt: 101.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1,II