2-[(3-Aminophenyl)thio]-4,6-dimethylpyrimidine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 3-[(4,6-Dimethylpyrimidin-2-yl)sulphanyl]aniline
OR6113-1G OR6113-5G OR6113-25G
APOSOR6113-1GEA 185 EUR
APOSOR6113-1G APOSOR6113-5G APOSOR6113-25G
2-[(3-Aminophenyl)thio]-4,6-dimethylpyrimidine 95+%
2-[(3-Aminophenyl)thio]-4,6-dimethylpyrimidine
Formula: C₁₂H₁₃N₃S
MW: 231.32 g/mol
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00203026
CAS Number: 387358-42-1