1-(4-Methoxyphenyl)cyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
Synonyms: 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid
OR61139-5G
APOSOR61139-5GEA 65.4 EUR
APOSOR61139-5G
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid 97%
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formula: C₁₁H₁₂O₃
MW: 192.2112 g/mol
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00019223
CAS Number: 16728-01-1