6-Aminonicotinamide 95+%
Supplier: Apollo Scientific
OR6120-10G OR6120-25G OR6120-5G
APOSOR6120-10GEA 275 EUR
APOSOR6120-10G APOSOR6120-25G APOSOR6120-5G
6-Aminonicotinamide 95+%
6-Aminonicotinamide
Formula: C₆H₇N₃O
Melting Pt: 245…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00006327
CAS Number: 329-89-5