2-Amino-4,6-dimethylpyridine 97%
Supplier: Apollo Scientific
Warning
OR6121-1G OR6121-25G OR6121-5G
APOSOR6121-1GEA 21.9 EUR
APOSOR6121-1G APOSOR6121-25G APOSOR6121-5G
2-Amino-4,6-dimethylpyridine 97%
2-Amino-4,6-dimethylpyridine
Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
Boiling Pt: 235 °C
Melting Pt: 63…64 °C
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006322
CAS Number: 5407-87-4
EINECS: 226-470-9