3-Amino-1H-pyrazole 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole
OR6125-100G OR6125-25G OR6125-5G
APOSOR6125-100GEA 72.5 EUR
APOSOR6125-100G APOSOR6125-25G APOSOR6125-5G
3-Amino-1H-pyrazole 97%
3-Amino-1H-pyrazole
Formula: C₃H₅N₃
MW: 83.09 g/mol
Boiling Pt: 218 °C/122mmHg
Melting Pt: 33…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
UN: 3263
ADR: 8,II