Dodecylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Dodecylamine, Dodecan-1-amine, 1-Aminododecane
OR61300-100G OR61300-1KG OR61300-500G
APOSOR61300-100GEA 79.9 EUR
APOSOR61300-100G APOSOR61300-1KG APOSOR61300-500G
Dodecylamine 97%
Dodecylamine
Formula: H₃C(CH₂)₁₀CH₂NH₂
MW: 185.35 g/mol
Boiling Pt: 247…249 °C
Melting Pt: 27…31 °C
Density: 0.806 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008154
CAS Number: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8 II