5-Isocyanato-1,3-benzodioxole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Benzodioxol-5-yl isocyanate, 5-Isocyanatobenzo[d][1,3]dioxole
OR61306-1G OR61306-5G
APOSOR61306-1GEA 55.1 EUR
APOSOR61306-1G APOSOR61306-5G
5-Isocyanato-1,3-benzodioxole 95%
5-Isocyanato-1,3-benzodioxole
When melted, may contain small amount of sediment.
Formula: C₈H₅NO₃
MW: 163.13 g/mol
Melting Pt: 48…51 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863692
CAS Number: 69922-28-7
UN: 3335
ADR: 9,III