6-Bromo-m-toluonitrile
Supplier: Apollo Scientific
Warning
OR61314-100G OR61314-25G OR61314-5G
APOSOR61314-100GEA 261 EUR
APOSOR61314-100G APOSOR61314-25G APOSOR61314-5G
6-Bromo-m-toluonitrile
6-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD11040281
CAS Number: 42872-83-3
UN: 3439
ADR: 6.1,III