5-Amino-1-ethylpyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 2-Ethyl-2H-pyrazol-3-ylamine, 1-ETHYL-PYRAZOL-5-AMINE, 1-Ethyl-1H-pyrazol-5-amine
OR61338-5G
APOSOR61338-5GEA 60.9 EUR
APOSOR61338-5G
5-Amino-1-ethylpyrazole
5-Amino-1-ethylpyrazole
Formula: C₅H₉N₃
MW: 111.148 g/mol
Boiling Pt: 92…94 °C/0.01mmHg
Melting Pt: 52…57 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00010849
CAS Number: 3528-58-3