6-Bromo-m-toluidine 98%
Supplier: Apollo Scientific
Warning
OR61400-100G OR61400-25G OR61400-500G
APOSOR61400-100GEA 66.7 EUR
APOSOR61400-100G APOSOR61400-25G APOSOR61400-500G
6-Bromo-m-toluidine 98%
6-Bromo-m-toluidine
Formula: C₇H₈BrN
Melting Pt: 32…34 °C
Density: 1.486 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01806278
CAS Number: 53078-85-6