1,4-Dioxane 99%
Supplier: Apollo Scientific
Danger
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
OR61424-1L OR61424-2.5L
APOSOR61424-1LEA 37.7 EUR
APOSOR61424-1L APOSOR61424-2.5L
1,4-Dioxane 99%
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 100…102 °C
Melting Pt: 44540 °C
Density: 1.034 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
UN: 1165
ADR: 3,II