4-Iodobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
OR61519-1G OR61519-25G OR61519-5G
APOSOR61519-1GEA 21.9 EUR
APOSOR61519-1G APOSOR61519-25G APOSOR61519-5G
4-Iodobenzaldehyde 98%
4-Iodobenzaldehyde
Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 265 °C
Melting Pt: 77…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00039576
CAS Number: 15164-44-0