2,4,6-Tribromopyridine 95%
Supplier: Apollo Scientific
Warning
OR61535-1G OR61535-5G
APOSOR61535-1GEA 77 EUR
APOSOR61535-1G APOSOR61535-5G
2,4,6-Tribromopyridine 95%
2,4,6-Tribromopyridine
Formula: C₅H₂Br₃N
MW: 315.7900 g/mol
Melting Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00234002
CAS Number: 2408-70-0