2,6-Diaminopurine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine
OR6157-1G OR6157-5G OR6157-10G OR6157-25G
APOSOR6157-1GEA 14.7 EUR
APOSOR6157-1G APOSOR6157-5G APOSOR6157-10G APOSOR6157-25G
2,6-Diaminopurine 98%
2,6-Diaminopurine
Formula: C₅H₆N₆
MW: 150.14 g/mol
Melting Pt: >300 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00213668
CAS Number: 1904-98-9
EINECS: 217-605-2