2,4-Xylenol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Hydroxy-3-xylene, 2,4-Dimethylphenol, 2,4-Xylenol
OR61605-100G OR61605-25G OR61605-500G
APOSOR61605-100GEA 50.9 EUR
APOSOR61605-100G APOSOR61605-25G APOSOR61605-500G
2,4-Xylenol 98%
2,4-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 209…213 °C
Melting Pt: 22…23 °C
Density: 1.02 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2928
ADR: 6.1+8,II