4-(Pyrimidin-2-yl)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
OR6161-1G OR6161-250MG
APOSOR6161-1GEA 280 EUR
APOSOR6161-1G APOSOR6161-250MG
4-(Pyrimidin-2-yl)benzaldehyde 95%
4-(Pyrimidin-2-yl)benzaldehyde
Formula: C₁₁H₈N₂O
Melting Pt: 108…109 °C
Storage Temperature: Ambient
MDL Number: MFCD08056282
CAS Number: 77232-38-3