2,3-Dichloro-4-hydroxybenzaldehyde 96%
Supplier: Apollo Scientific
Warning
OR6167-1G OR6167-250MG OR6167-5G
APOSOR6167-1GEA 216 EUR
APOSOR6167-1G APOSOR6167-250MG APOSOR6167-5G
2,3-Dichloro-4-hydroxybenzaldehyde 96%
2,3-Dichloro-4-hydroxybenzaldehyde
Formula: C₇H₄Cl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07775189
CAS Number: 16861-22-6