2-Bromo-3-formylpyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde
OR6170-25G OR6170-5G OR6170-100G
APOSOR6170-25GEA 143 EUR
APOSOR6170-25G APOSOR6170-5G APOSOR6170-100G
2-Bromo-3-formylpyridine 98%
2-Bromo-3-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Boiling Pt: 100 °C/3mmHg
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD04966945
CAS Number: 128071-75-0