2-Chloro-4-formylpyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde
OR6172-1G OR6172-5G
APOSOR6172-1GEA 21.9 EUR
APOSOR6172-1G APOSOR6172-5G
2-Chloro-4-formylpyridine 97%
2-Chloro-4-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06651557
CAS Number: 101066-61-9