2-Chloro-5-formylpyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Chloronicotinaldehyde, 2-Chloropyridine-5-carboxaldehyde, 6-Chloropyridine-3-carboxaldehyde
OR6178-1G OR6178-5G OR6178-10G
APOSOR6178-1GEA 21.9 EUR
APOSOR6178-1G APOSOR6178-5G APOSOR6178-10G
2-Chloro-5-formylpyridine 95%
2-Chloro-5-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03095223
CAS Number: 23100-12-1