4,4'-Dibromobenzophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: Bis(4-bromophenyl) ketone
OR62073-1G OR62073-5G OR62073-25G OR62073-100G OR62073-500G
APOSOR62073-1GEA 14.7 EUR
APOSOR62073-1G APOSOR62073-5G APOSOR62073-25G APOSOR62073-100G APOSOR62073-500G
4,4'-Dibromobenzophenone 98%
4,4'-Dibromobenzophenone
Formula: C₁₃H₈Br₂O
MW: 340.01 g/mol
Melting Pt: 171…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00016330
CAS Number: 3988-03-2
EINECS: 223-632-0