(3-Methylenecyclobutyl)methylamine 95%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Aminomethyl)-3-methylenecyclobutane
OR6220-1G OR6220-250MG OR6220-50MG OR6220-500MG
APOSOR6220-1GEA 434 EUR
APOSOR6220-1G APOSOR6220-250MG APOSOR6220-50MG APOSOR6220-500MG
(3-Methylenecyclobutyl)methylamine 95%
(3-Methylenecyclobutyl)methylamine
Formula: C₆H₁₁N
Boiling Pt: 132…133 °C/571mmHg
Storage Temperature: Ambient
MDL Number: MFCD08235202
CAS Number: 16333-93-0
UN: 1760
ADR: 8,II