N-(4,6-Dimethylpyrimidin-2-yl)-4-aminobenzoic acid 95+%
Supplier: Apollo Scientific
OR6225-1G OR6225-5G OR6225-25G
APOSOR6225-1GEA 185 EUR
APOSOR6225-1G APOSOR6225-5G APOSOR6225-25G
N-(4,6-Dimethylpyrimidin-2-yl)-4-aminobenzoic acid 95+%
N-(4,6-Dimethylpyrimidin-2-yl)-4-aminobenzoic acid
Formula: C₁₃H₁₃N₃O₂
MW: 243.27 g/mol
Melting Pt: >335 °C
Storage Temperature: Ambient
MDL Number: MFCD00203264
CAS Number: 81261-97-4