4-Bromo-m-toluonitrile
Supplier: Apollo Scientific
Warning
OR6236-25G OR6236-5G
APOSOR6236-25GEA 85.7 EUR
APOSOR6236-25G APOSOR6236-5G
4-Bromo-m-toluonitrile
4-Bromo-m-toluonitrile
Formula: C₈H₆BrN
Boiling Pt: 129…130 °C/5mmHg
Melting Pt: 53…57 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00031538
CAS Number: 41963-20-6