4-Methylpicolinic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methylpicolinic acid, 4-Methylpyridine-2-carboxylic acid
OR6256-1G OR6256-5G
APOSOR6256-1GEA 34.8 EUR
APOSOR6256-1G APOSOR6256-5G
4-Methylpicolinic acid 97%
4-Methylpicolinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD03731175
CAS Number: 4021-08-3