4,5-Dimethylthiophene-2-carboxaldehyde
Supplier: Apollo Scientific
Warning
OR63020-1G OR63020-25G OR63020-5G
APOSOR63020-1GEA 168 EUR
APOSOR63020-1G APOSOR63020-25G APOSOR63020-5G
4,5-Dimethylthiophene-2-carboxaldehyde
4,5-Dimethylthiophene-2-carboxaldehyde
Formula: C₇H₈OS
MW: 140.2 g/mol
Boiling Pt: 82…85 °C
Density: 1.179 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093897
CAS Number: 5928-48-3