3-Chloro-6-methylpyridazine-4-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
OR6303-1G OR6303-5G
APOSOR6303-1GEA 357 EUR
APOSOR6303-1G APOSOR6303-5G
3-Chloro-6-methylpyridazine-4-carboxylic acid 97%
3-Chloro-6-methylpyridazine-4-carboxylic acid
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD08235214
CAS Number: 914637-40-4