1,1-Cyclopropanedicarboxylic acid 98%
Supplier: Apollo Scientific
Danger
OR6347-100G OR6347-25G
APOSOR6347-100GEA 78.3 EUR
APOSOR6347-100G APOSOR6347-25G
1,1-Cyclopropanedicarboxylic acid 98%
1,1-Cyclopropanedicarboxylic acid
Formula: C₅H₆O₄
MW: 130.1 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013727
CAS Number: 598-10-7
EINECS: 209-917-2
UN: 3261
ADR: 8,III