2,3-Dichlorophenol 98+%
Supplier: Apollo Scientific
Warning
OR6358-100G OR6358-25G OR6358-500G
APOSOR6358-100GEA 84.4 EUR
APOSOR6358-100G APOSOR6358-25G APOSOR6358-500G
2,3-Dichlorophenol 98+%
2,3-Dichlorophenol
Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 206 °C
Melting Pt: 57…60 °C
Density: 1.4
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002160
CAS Number: 576-24-9
UN: 2020
ADR: 6.1 III