3-Chlorophenylhydrazinium chloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Chlorophenyl)hydrazine hydrochloride, 3-Chlorophenylhydrazine hydrochloride
OR6387-100G OR6387-250G OR6387-25G
APOSOR6387-100GEA 30.6 EUR
APOSOR6387-100G APOSOR6387-250G APOSOR6387-25G
3-Chlorophenylhydrazinium chloride 97%
3-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
Melting Pt: 242 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02656655
CAS Number: 2312-23-4
EINECS: 219-005-6
UN: 2811
ADR: 6.1,III