Methyl-6-methylnicotinate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 6-methylnicotinate
OR6396-100G OR6396-25G
APOSOR6396-100GEA 87 EUR
APOSOR6396-100G APOSOR6396-25G
Methyl-6-methylnicotinate 95%
Methyl 6-methylnicotinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 152…154 °C/40mmHg
Melting Pt: <30 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006340
CAS Number: 5470-70-2