2,5-Dibromotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Dibromo-2-methylbenzene
OR6405-100G OR6405-500G
APOSOR6405-100GEA 88.6 EUR
APOSOR6405-100G APOSOR6405-500G
2,5-Dibromotoluene 98%
2,5-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 135…136 °C
Melting Pt: 44352 °C
Density: 1.815 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000090
CAS Number: 615-59-8