4-Amino-3-chloropyridine 97%
Supplier: Apollo Scientific
Warning
OR6413-5G OR6413-1G
APOSOR6413-5GEA 24.8 EUR
APOSOR6413-5G APOSOR6413-1G
4-Amino-3-chloropyridine 97%
4-Amino-3-chloropyridine
Formula: C₅H₅ClN₂
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD03984473
CAS Number: 19798-77-7