4-Chloro-2-(methylthio)pyrimidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Chloro-2-methylthiopyrimidine
OR6414-100G OR6414-25G OR6414-5G
APOSOR6414-100GEA 190 EUR
APOSOR6414-100G APOSOR6414-25G APOSOR6414-5G
4-Chloro-2-(methylthio)pyrimidine 97%
4-Chloro-2-(methylthio)pyrimidine
Formula: C₅H₅ClN₂S
Boiling Pt: 139…140 °C/36mmHg
Melting Pt: –2 °C
Density: 1.381 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00006083
CAS Number: 49844-90-8
EINECS: 256-500-6
UN: 1760
ADR: 8,II