1,1-Bis(hydroxymethyl)cyclopropane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1-Bis(Hydroxymethyl)cyclopropane
OR6432-100G OR6432-25G OR6432-500G
APOSOR6432-100GEA 40.6 EUR
APOSOR6432-100G APOSOR6432-25G APOSOR6432-500G
1,1-Bis(hydroxymethyl)cyclopropane 98%
1,1-Bis(hydroxymethyl)cyclopropane
Formula: C₅H₁₀O₂
MW: 102.133 g/mol
Boiling Pt: 235…236 °C
Density: 1.065 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00464207
CAS Number: 39590-81-3