3-Methoxybenzyl bromide 98%
Supplier: Apollo Scientific
|
Danger
|
OR6438-100G
OR6438-25G
OR6438-5G
APOSOR6438-100GEA
394
EUR
APOSOR6438-100G
APOSOR6438-25G
APOSOR6438-5G
3-Methoxybenzyl bromide 98%
3-Methoxybenzyl bromide
Formula:
C₈H₉BrO Boiling Pt: 152 °C Density: 1.436 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216590 CAS Number: 874-98-6 UN: 3265 ADR: 8,II |