α-Bromo-m-xylene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Methylbenzyl bromide
OR6442-100G OR6442-25G OR6442-5G
APOSOR6442-100GEA 261 EUR
APOSOR6442-100G APOSOR6442-25G APOSOR6442-5G
α-Bromo-m-xylene 98%
α-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 184…185 °C/340mmHg
Density: 1.37 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000177
CAS Number: 620-13-3
EINECS: 210-625-2
UN: 1701
ADR: 6.1,II