N-(4-Aminophenyl)-4,6-dimethyl-2-pyrimidinamine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: N1-(4,6-Dimethylpyrimidin-2-yl)benzene-1,4-diamine
OR6449-1G OR6449-100MG
APOSOR6449-1GEA 429 EUR
APOSOR6449-1G APOSOR6449-100MG
N-(4-Aminophenyl)-4,6-dimethyl-2-pyrimidinamine 95+%
N-(4-Aminophenyl)-4,6-dimethyl-2-pyrimidinamine
Formula: C₁₂H₁₄N₄
MW: 214.27 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00202977
CAS Number: 81261-93-0