4-Isopropylanisole 98%
Supplier: Apollo Scientific
Warning
OR6453-100G OR6453-25G
APOSOR6453-100GEA 87 EUR
APOSOR6453-100G APOSOR6453-25G
4-Isopropylanisole 98%
4-Isopropylanisole
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 212…213 °C
Density: 0.935 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00015033
CAS Number: 4132-48-3
EINECS: 223-952-0