(Methylamino)acetaldehyde dimethylacetal 97%
Supplier: Apollo Scientific
Warning
OR6502-100G OR6502-25G
APOSOR6502-100GEA 94.5 EUR
APOSOR6502-100G APOSOR6502-25G
(Methylamino)acetaldehyde dimethylacetal 97%
(Methylamino)acetaldehyde dimethylacetal
Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 139…140 °C
Melting Pt: –73 °C
Density: 0.928 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00008485
CAS Number: 122-07-6
EINECS: 204-520-0
UN: 1993
ADR: 3,III