(±)-1-Indanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (±)-1-Hydroxyindan, 1-Indanol, 2,3-Dihydro-1H-inden-1-ol
OR6504-100G OR6504-25G OR6504-5G
APOSOR6504-100GEA 59.6 EUR
APOSOR6504-100G APOSOR6504-25G APOSOR6504-5G
(±)-1-Indanol 98%
(±)-1-Indanol
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 128 °C/12mmHg
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00003797
CAS Number: 6351-10-6