6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
Supplier: Apollo Scientific
Warning
OR6513-100G OR6513-25G OR6513-5G
APOSOR6513-100GEA 95.7 EUR
APOSOR6513-100G APOSOR6513-25G APOSOR6513-5G
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₁₁H₁₆ClNO₂
MW: 229.71 g/mol
Melting Pt: 260…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00012744
CAS Number: 2328-12-3