Thiomorpholine 98%
Supplier: Apollo Scientific
Danger
OR6523-25G OR6523-5G
APOSOR6523-25GEA 34.8 EUR
APOSOR6523-25G APOSOR6523-5G
Thiomorpholine 98%
Thiomorpholine
Formula: C₄H₉NS
MW: 103.19 g/mol
Boiling Pt: 169…177 °C
Density: 1.026 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005974
CAS Number: 123-90-0
EINECS: 204-660-2
UN: 3267
ADR: 8 III