3-Amino-2-chloropyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine
OR6543-25G OR6543-5G OR6543-1G OR6543-500G
APOSOR6543-25GEA 21.9 EUR
APOSOR6543-25G APOSOR6543-5G APOSOR6543-1G APOSOR6543-500G
3-Amino-2-chloropyridine 98%
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 130…134 °C/22mmHg
Melting Pt: 76…78 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9