N-(4,6-Dimethylpyrimidin-2-yl)-3-aminobenzoic acid 95+%
Supplier: Apollo Scientific
Warning
OR6555-1G OR6555-250MG
APOSOR6555-1GEA 229 EUR
APOSOR6555-1G APOSOR6555-250MG
N-(4,6-Dimethylpyrimidin-2-yl)-3-aminobenzoic acid 95+%
N-(4,6-Dimethylpyrimidin-2-yl)-3-aminobenzoic acid
Formula: C₁₃H₁₃N₃O₂
MW: 243.27 g/mol
Melting Pt: >335 °C
Storage Temperature: Ambient
MDL Number: MFCD00203262
CAS Number: 81261-77-0