Diethyl mesoxalate (diethyl ketomalonate) 97%
Supplier: Apollo Scientific
Warning
Synonyms: Diethyl 2-oxomalonate
OR6568-25G OR6568-5G OR6568-100G
APOSOR6568-25GEA 33.5 EUR
APOSOR6568-25G APOSOR6568-5G APOSOR6568-100G
Diethyl mesoxalate (diethyl ketomalonate) 97%
Diethyl mesoxalate (Diethyl ketomalonate)
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 208…210 °C
Density: 1.142 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009121
CAS Number: 609-09-6
EINECS: 210-176-2