2-Bromothiophene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Thienyl bromide
OR6569-1KG OR6569-500G
APOSOR6569-1KGEA 275 EUR
APOSOR6569-1KG APOSOR6569-500G
2-Bromothiophene 98%
2-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 149…151 °C
Melting Pt: < –10 °C
Density: 1.684 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005417
CAS Number: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1+3 II